Assay Details
Gene Symbol: KCNJ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004004689.4 | XP_004004738.2 | 1 - 2 | 1217 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
