Assay Details
| Gene Symbol: GPR180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973708.1 | XP_027829509.1 | 4 - 5 | 750 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GPR180| Gene Symbol: GPR180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973708.1 | XP_027829509.1 | 4 - 5 | 750 | 85 | |
Set Membership: |
|