Assay Details
Gene Symbol: HJV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015092214.2 | XP_014947700.2 | 2 - 3 | 364 | 81 | |
XM_015092215.2 | XP_014947701.2 | 2 - 3 | 383 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
