Assay Details
Gene Symbol: ASB7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004017712.4 | XP_004017761.1 | 4 - 5 | 605 | 84 | |
XM_012098320.3 | XP_011953710.1 | 4 - 5 | 616 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
