Assay Details
| Gene Symbol: ZNF384 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004023275.4 | XP_004023324.2 | 8 - 9 | 1984 | 71 | |
| XM_027967945.1 | XP_027823746.1 | 11 - 12 | 2158 | 71 | ||
| XM_027967946.1 | XP_027823747.1 | 11 - 12 | 2298 | 71 | ||
| XM_027967947.1 | XP_027823748.1 | 11 - 12 | 1753 | 71 | ||
| XM_027967948.1 | XP_027823749.1 | 9 - 10 | 2213 | 71 | ||
| XM_027967949.1 | XP_027823750.1 | 9 - 10 | 2167 | 71 | ||
| XM_027967950.1 | XP_027823751.1 | 8 - 9 | 2119 | 71 | ||
| XM_027967951.1 | XP_027823752.1 | 9 - 10 | 2079 | 71 | ||
More Information
Set Membership: |
|

ZNF384
Genomic Map