Assay Details
Gene Symbol: DDX3X | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004022011.3 | XP_004022060.1 | 5 - 6 | 540 | 62 | |
XM_012106516.2 | XP_011961906.1 | 5 - 6 | 540 | 62 | ||
XM_012106517.2 | XP_011961907.1 | 5 - 6 | 537 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
