Assay Details
Gene Symbol: LOC101122444 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004011715.4 | XP_004011764.2 | 3 - 4 | 393 | 87 | |
XM_012183791.3 | XP_012039181.1 | 3 - 4 | 379 | 87 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
