Assay Details
| Gene Symbol: JDP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004011077.4 | XP_004011126.1 | 3 - 4 | 2270 | 67 | |
| XM_012182070.3 | XP_012037460.1 | 3 - 4 | 510 | 67 | ||
| XM_015097184.2 | XP_014952670.1 | 3 - 4 | 440 | 67 | ||
| XM_027972041.1 | XP_027827842.1 | 3 - 4 | 428 | 67 | ||
More Information
Set Membership: |
|

JDP2
Genomic Map