Assay Details
Gene Symbol: DDIT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004006542.4 | XP_004006591.1 | 3 - 4 | 311 | 89 | |
GenBank mRNA | AY943948.1 | - | 1 - 2 | 70 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
