Assay Details
Gene Symbol: SST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009196.1 | NP_001009196.1 | 1 - 2 | 172 | 86 | |
GenBank mRNA | AF031488.1 | - | 1 - 2 | 172 | 86 | |
Y15267.1 | - | 172 | 86 |
Gene Symbol: LOC114113840 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027966037.1 | XP_027821838.1 | 1 - 2 | 233 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
