Assay Details
| Gene Symbol: GHR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009323.2 | NP_001009323.1 | 2 - 3 | 261 | 70 | |
| XM_012096676.2 | XP_011952066.1 | 1 - 2 | 89 | 70 | ||
| XM_012096677.3 | XP_011952067.1 | 2 - 3 | 143 | 70 | ||
| XM_027979937.1 | XP_027835738.1 | 2 - 3 | 517 | 70 | ||
| GenBank mRNA | M82912.1 | - | 2 - 3 | 263 | 70 | |
More Information
Set Membership: |
|

GHR
Genomic Map