Assay Details
Gene Symbol: LOC101109747 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001280717.1 | NP_001267646.1 | 2 - 3 | 326 | 57 | |
XM_012100429.3 | XP_011955819.2 | 2 - 3 | 414 | 57 | ||
GenBank mRNA | FM986335.1 | - | 2 - 3 | 329 | 57 | |
FM986336.1 | - | 2 - 3 | 329 | 57 | ||
FM986337.1 | - | 2 - 3 | 329 | 57 | ||
HQ848644.1 | - | 2 - 3 | 287 | 57 | ||
HQ848645.1 | - | 2 - 3 | 287 | 57 | ||
HQ848646.1 | - | 2 - 3 | 287 | 57 | ||
JF735994.1 | - | 2 - 3 | 326 | 57 | ||
JQ824376.1 | - | 2 - 3 | 354 | 57 | ||
M73983.1 | - | 2 - 3 | 351 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
