Assay Details
Gene Symbol: TIMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009319.2 | NP_001009319.1 | 2 - 3 | 270 | 55 | |
GenBank mRNA | S67450.1 | - | 2 - 3 | 270 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
