Assay Details
Gene Symbol: RAB8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161860.1 | NP_001155332.1 | 4 - 5 | 327 | 64 | |
GenBank mRNA | FJ943968.1 | - | 4 - 5 | 327 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
