Assay Details
Gene Symbol: REN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009299.1 | NP_001009299.1 | 8 - 9 | 994 | 59 | |
GenBank mRNA | L43524.1 | - | 8 - 9 | 994 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
