Assay Details
| Gene Symbol: THRA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100919.3 | NP_001094389.1 | 5 - 6 | 885 | 62 | |
| XM_027973954.1 | XP_027829755.1 | 6 - 7 | 1075 | 62 | ||
| GenBank mRNA | Z68308.1 | - | 6 - 7 | 1097 | 62 | |
More Information
Set Membership: |
|

THRA
Genomic Map