Assay Details
| Gene Symbol: CCL28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129903.1 | NP_001123375.1 | 2 - 3 | 226 | 89 | |
| GenBank mRNA | DQ181939.1 | - | 1 - 2 | 120 | 89 | |
| EF624458.1 | - | 2 - 3 | 226 | 89 | ||
| EU294303.1 | - | 1 - 2 | 97 | 89 | ||
More Information
Set Membership: |
|

CCL28
Genomic Map