Assay Details
| Gene Symbol: TUBA4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163055.2 | NP_001156527.1 | 2 - 3 | 279 | 87 | |
| GenBank mRNA | EE753586.1 | - | 2 - 3 | 279 | 87 | |
| EE767677.1 | - | 2 - 3 | 272 | 87 | ||
| FJ958352.1 | - | 1 - 2 | 227 | 87 | ||
More Information
Set Membership: |
|

TUBA4A
Genomic Map