Assay Details
| Gene Symbol: TIMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166186.1 | NP_001159658.1 | 4 - 5 | 469 | 87 | |
| GenBank mRNA | FJ384971.1 | - | 3 - 4 | 285 | 87 | |
| GQ221040.1 | - | 4 - 5 | 469 | 87 | ||
More Information
Set Membership: |
|

TIMP2
Genomic Map