Assay Details
| Gene Symbol: IFI30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205150.1 | NP_001192079.1 | 4 - 5 | 468 | 70 | |
| GenBank mRNA | HM017967.1 | - | 4 - 5 | 468 | 70 | |
More Information
Set Membership: |
|

IFI30
Genomic Map