Assay Details
| Gene Symbol: UCP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001308581.1 | NP_001295510.1 | 2 - 3 | 313 | 56 | |
| GenBank mRNA | KF015729.1 | - | 2 - 3 | 313 | 56 | |
More Information
Set Membership: |
|

UCP3
Genomic Map