Assay Details
Gene Symbol: PTGDS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009257.2 | NP_001009257.1 | 5 - 6 | 581 | 71 | |
GenBank mRNA | AJ133642.1 | - | 5 - 6 | 575 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
