Assay Details
Gene Symbol: NAGA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126345.1 | NP_001119817.1 | 2 - 3 | 150 | 71 | |
XM_012158583.2 | XP_012013973.1 | 3 - 4 | 71 | |||
XM_012158598.2 | XP_012013988.1 | 3 - 4 | 71 | |||
GenBank mRNA | EU579460.1 | - | 2 - 3 | 150 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
