Assay Details
| Gene Symbol: ATP5PO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001165889.2 | NP_001159361.1 | 2 - 3 | 124 | 63 | |
| GenBank mRNA | FJ546085.1 | - | 2 - 3 | 125 | 63 | |
More Information
Set Membership: |
|

ATP5PO
Genomic Map