Assay Details
| Gene Symbol: LIN28B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256768.1 | NP_001243697.1 | 2 - 3 | 234 | 85 | |
| XM_012182375.3 | XP_012037765.1 | 2 - 3 | 234 | 85 | ||
| XM_015097249.2 | XP_014952735.1 | 1 - 2 | 197 | 85 | ||
| GenBank mRNA | JQ277700.1 | - | 2 - 3 | 234 | 85 | |
| JQ277701.1 | - | 2 - 3 | 234 | 85 | ||
More Information
Set Membership: |
|

LIN28B
Genomic Map