Assay Details
Gene Symbol: CIDEC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306115.1 | NP_001293044.1 | 4 - 5 | 655 | 87 | |
XM_004018278.3 | XP_004018327.1 | 4 - 5 | 1552 | 87 | ||
XM_012099534.3 | XP_011954924.1 | 4 - 5 | 812 | 87 | ||
XM_012099535.3 | XP_011954925.1 | 4 - 5 | 405 | 87 | ||
XM_027957552.1 | XP_027813353.1 | 2 - 3 | 234 | 87 | ||
GenBank mRNA | KM199684.1 | - | 4 - 5 | 655 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
