Assay Details
| Gene Symbol: KISS1R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318077.1 | NP_001305006.1 | 2 - 3 | 369 | 93 | |
| GenBank mRNA | EU272411.1 | - | 2 - 3 | 264 | 93 | |
| EU272412.1 | - | 2 - 3 | 153 | 93 | ||
| JQ812137.1 | - | 2 - 3 | 369 | 93 | ||
More Information
Set Membership: |
|

KISS1R
Genomic Map