Assay Details
| Gene Symbol: KISS1R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318077.1 | NP_001305006.1 | 3 - 4 | 505 | 74 | |
| GenBank mRNA | EU272411.1 | - | 3 - 4 | 400 | 74 | |
| EU272412.1 | - | 3 - 4 | 289 | 74 | ||
| JQ812137.1 | - | 3 - 4 | 505 | 74 | ||
More Information
Set Membership: |
|

KISS1R
Genomic Map