Assay Details
| Gene Symbol: ADORA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009775.2 | NP_001009775.2 | 1 - 2 | 562 | 71 | |
| GenBank mRNA | DY518446.1 | - | 1 - 2 | 529 | 71 | |
| S65334.1 | - | 1 - 2 | 575 | 71 | ||
More Information
Set Membership: |
|

ADORA3
Genomic Map