Assay Details
Gene Symbol: RPS17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001280688.1 | NP_001267617.1 | 1 - 2 | 173 | 97 | |
GenBank mRNA | JX534531.1 | - | 1 - 2 | 173 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
