Assay Details
Gene Symbol: REM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161859.1 | NP_001155331.1 | 2 - 3 | 425 | 85 | |
XM_012188065.2 | XP_012043455.1 | 3 - 4 | 582 | 85 | ||
GenBank mRNA | FJ943967.1 | - | 2 - 3 | 425 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
