Assay Details
Gene Symbol: CST3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001280713.1 | NP_001267642.1 | 2 - 3 | 412 | 71 | |
GenBank mRNA | JX534543.1 | - | 2 - 3 | 412 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
