Assay Details
| Gene Symbol: KRT71 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001280716.1 | NP_001267645.1 | 4 - 5 | 886 | 67 | |
| GenBank mRNA | JX975451.1 | - | 4 - 5 | 886 | 67 | |
More Information
Set Membership: |
|

KRT71
Genomic Map