Assay Details
Gene Symbol: BCAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009444.1 | NP_001009444.1 | 3 - 4 | 764 | 61 | |
XM_012159865.3 | XP_012015255.2 | 3 - 4 | 381 | 61 | ||
GenBank mRNA | AF184916.1 | - | 3 - 4 | 764 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
