Assay Details
Gene Symbol: MT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009755.1 | NP_001009755.1 | 1 - 2 | 33 | 90 | |
GenBank mRNA | AF500199.1 | - | 1 - 2 | 33 | 90 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: MT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009755.1 | NP_001009755.1 | 1 - 2 | 33 | 90 | |
GenBank mRNA | AF500199.1 | - | 1 - 2 | 33 | 90 |