Assay Details
Gene Symbol: MT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009755.1 | NP_001009755.1 | 2 - 3 | 96 | 84 | |
GenBank mRNA | AF500199.1 | - | 2 - 3 | 96 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
