Assay Details
Gene Symbol: BGLAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040009.2 | NP_001035098.1 | 1 - 2 | 69 | 57 | |
GenBank mRNA | DQ418490.1 | - | 1 - 2 | 73 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() |
