Assay Details
| Gene Symbol: GKN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099630.1 | NP_001093100.1 | 4 - 5 | 324 | 67 | |
| GenBank mRNA | EF617338.1 | - | 4 - 5 | 324 | 67 | |
More Information
Set Membership: |
|

GKN1
Genomic Map