Assay Details
| Gene Symbol: GKN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099630.1 | NP_001093100.1 | 5 - 6 | 471 | 72 | |
| GenBank mRNA | EF617338.1 | - | 5 - 6 | 471 | 72 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: GKN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099630.1 | NP_001093100.1 | 5 - 6 | 471 | 72 | |
| GenBank mRNA | EF617338.1 | - | 5 - 6 | 471 | 72 | |