Assay Details
| Gene Symbol: PPIL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163049.2 | NP_001156521.1 | 4 - 5 | 576 | 80 | |
| XM_027963867.1 | XP_027819668.1 | 5 - 6 | 597 | 80 | ||
| XM_027963868.1 | XP_027819669.1 | 5 - 6 | 358 | 80 | ||
| GenBank mRNA | EE812467.1 | - | 4 - 5 | 295 | 80 | |
| FJ958362.1 | - | 2 - 3 | 172 | 80 | ||
More Information
Set Membership: |
|

PPIL3
Genomic Map