Assay Details
| Gene Symbol: NFKBIA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166184.1 | NP_001159656.1 | 5 - 6 | 997 | 63 | |
| GenBank mRNA | EU366482.1 | - | 2 - 3 | 359 | 63 | |
| GQ221037.1 | - | 5 - 6 | 997 | 63 | ||
More Information
Set Membership: |
|

NFKBIA
Genomic Map