Assay Details
Gene Symbol: AKIRIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252176.1 | NP_001239105.1 | 2 - 3 | 379 | 96 | |
GenBank mRNA | JN390431.1 | - | 2 - 3 | 379 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() |
