Assay Details
| Gene Symbol: ARF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162548.1 | NP_001156020.1 | 1 | 324 | 92 | |
| XM_015096683.2 | XP_014952169.1 | 2 | 873 | 92 | ||
| XM_015096684.2 | XP_014952170.1 | 2 | 1345 | 92 | ||
| GenBank mRNA | FJ969411.1 | - | 1 | 324 | 92 | |
More Information
Set Membership: |
|

ARF6
Genomic Map