Assay Details
| Gene Symbol: ABHD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104933.1 | NP_001098403.1 | 4 - 5 | 718 | 62 | |
| GenBank mRNA | EF660331.1 | - | 4 - 5 | 718 | 62 | |
More Information
Set Membership: |
|

ABHD5
Genomic Map