Assay Details
Gene Symbol: SLC25A15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127276.1 | NP_001120748.1 | 2 - 3 | 315 | 85 | |
XM_015097954.2 | XP_014953440.1 | 3 - 4 | 85 | |||
GenBank mRNA | EU626212.1 | - | 2 - 3 | 315 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
