Assay Details
| Gene Symbol: SLC39A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163601.1 | NP_001157073.1 | 3 - 4 | 301 | 80 | |
| XM_012180584.2 | XP_012035974.1 | 3 - 4 | 898 | 80 | ||
| GenBank mRNA | FJ937951.1 | - | 3 - 4 | 301 | 80 | |
More Information
Set Membership: |
|

SLC39A2
Genomic Map