Assay Details
Gene Symbol: PRP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112819.1 | NP_001106290.1 | 4 - 5 | 572 | 98 | |
GenBank mRNA | AB231298.1 | - | 4 - 5 | 572 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
