Assay Details
Gene Symbol: AMH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001308599.1 | NP_001295528.1 | 1 - 2 | 468 | 66 | |
GenBank mRNA | KC986978.1 | - | 1 - 2 | 468 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
