Assay Details
| Gene Symbol: AQP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009194.1 | NP_001009194.1 | 1 - 2 | 392 | 67 | |
| GenBank mRNA | AF009037.1 | - | 1 - 2 | 392 | 67 | |
More Information
Set Membership: |
|

AQP1
Genomic Map