Assay Details
| Gene Symbol: GUCA2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105261.1 | NP_001098731.1 | 1 - 2 | 78 | 68 | |
| GenBank mRNA | EF654536.1 | - | 1 - 2 | 78 | 68 | |
More Information
Set Membership: |
|

GUCA2A
Genomic Map